Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014584.2 | NP_055399.1 | 1, 2 | 572 | ||
XM_011536684.2 | XP_011534986.1 | 1, 2 | 391 | |||
GenBank mRNA | AF081886.1 | - | 1, 2 | 348 | ||
AK292839.1 | - | 1, 2 | 170 | |||
AY358463.1 | - | 1, 2 | 294 | |||
BC008674.1 | - | 1, 2 | 344 | 3866930 | ||
BC012941.1 | - | 1, 2 | 355 | 3868538 | ||
BX420309.2 | - | 1, 2 | 577 | |||
DR006292.1 | - | 1, 2 | 494 |