Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014597.4 | NP_055412.2 | 2 | 333 | ||
GenBank mRNA | AB046574.1 | - | 2 | 313 | ||
AK222722.1 | - | 2 | 306 | |||
AK292379.1 | - | 2 | 306 | |||
AK302909.1 | - | 2 | 289 | |||
AU134239.1 | - | 2 | 333 | |||
AY336729.1 | - | 2 | 314 | |||
AY394925.1 | - | 2 | 299 | |||
BC001784.1 | - | 2 | 308 | |||
BC020634.1 | - | 2 | 313 | |||
BC047688.1 | - | 2 | 295 | |||
BC130622.1 | - | 2 | 311 | 40146653 | ||
U15552.1 | - | 2 | 299 |