Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182577.2 | NP_872383.1 | 4 | 832 | ||
XM_005259545.4 | XP_005259602.1 | 4 | 1005 | |||
XM_017026671.1 | XP_016882160.1 | 3 | 402 | |||
GenBank mRNA | AK097378.1 | - | 4 | 832 | ||
BC104468.1 | - | 3 | 621 | 40032428 | ||
BC104469.1 | - | 4 | 725 | 40032432 |