Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014617.3 | NP_055432.2 | 3 | 297 | ||
GenBank mRNA | BC114456.1 | - | 2 | 396 | 40014816 | |
BM698524.1 | - | 3 | 297 | |||
EF426311.1 | - | 3 | 289 | |||
KT001516.1 | - | 3 | 299 |