Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284260.1 | NP_001271189.1 | 6 | 2046 | ||
NM_001284261.1 | NP_001271190.1 | 5 | 1347 | |||
NM_001284262.1 | NP_001271191.1 | 6 | 1483 | |||
NM_198285.2 | NP_938026.2 | 6 | 1982 | |||
XM_005249989.3 | XP_005250046.1 | 5 | 1843 | |||
XM_011516148.1 | XP_011514450.1 | 6 | 1531 | |||
XM_011516150.1 | XP_011514452.1 | 7 | 1735 | |||
GenBank mRNA | AK125347.1 | - | 5 | 1296 | ||
AK296049.1 | - | 6 | 1639 | |||
BC107125.2 | - | 5 | 1379 | 40023951 |