Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198540.2 | NP_940942.1 | 3 | 1610 | ||
XM_011526934.2 | XP_011525236.1 | 2 | 2929 | |||
GenBank mRNA | AB175895.1 | - | 1 | 1156 | ||
AJ419172.1 | - | 1 | 1156 | |||
AY277592.2 | - | 3 | 1610 |