Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014640.4 | NP_055455.3 | 3 | 861 | ||
XM_006712875.2 | XP_006712938.1 | 3 | 871 | |||
XM_017005387.1 | XP_016860876.1 | 3 | 871 | |||
XM_017005388.1 | XP_016860877.1 | 4 | 758 | |||
XM_017005389.1 | XP_016860878.1 | 4 | 777 | |||
XM_017005390.1 | XP_016860879.1 | 3 | 711 | |||
XM_017005391.1 | XP_016860880.1 | 2 | 776 | |||
XM_017005392.1 | XP_016860881.1 | 1 | 248 | |||
GenBank mRNA | AK291770.1 | - | 3 | 819 | ||
AK302728.1 | - | 3 | 407 | |||
AL833511.1 | - | 4 | 702 | |||
BC021707.2 | - | 3 | 786 | 4824604 | ||
D79995.1 | - | 3 | 698 |