Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301339.1 | NP_001288268.1 | 4 | 628 | ||
NM_213720.2 | NP_998885.1 | 4 | 607 | |||
NR_125755.1 | 4 | 554 | ||||
NR_125756.1 | 3 | 387 | ||||
GenBank mRNA | BC053570.1 | - | 4 | 558 | ||
BC065232.1 | - | 4 | 589 | 5747937 |