Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301339.1 | NP_001288268.1 | 3 | 517 | ||
NM_213720.2 | NP_998885.1 | 3 | 496 | |||
NR_125755.1 | 3 | 443 | ||||
NR_125756.1 | 2 | 276 | ||||
GenBank mRNA | AK289560.1 | - | 3 | 475 | ||
BC053570.1 | - | 3 | 447 | |||
BC065232.1 | - | 3 | 478 | 5747937 | ||
CN273854.1 | - | 3 | 391 |