Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214711.3 | NP_999876.2 | 5 | 2968 | ||
GenBank mRNA | BX640892.1 | - | 5 | 2879 | ||
BX640924.1 | - | 5 | 2920 | |||
BX640997.1 | - | 5 | 2955 | |||
BX647183.1 | - | 6 | 2787 | |||
BX647185.1 | - | 7 | 2897 | |||
BX648724.1 | - | 5 | 2912 | |||
BX648872.1 | - | 5 | 2840 | |||
BX648895.1 | - | 5 | 2918 | |||
BX648896.1 | - | 6 | 2852 |