Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001343.3 | NP_001001343.2 | 2 | 1384 | ||
GenBank mRNA | BC022570.1 | - | 2 | 1338 | 4793169 |