Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004558.3 | NP_004549.1 | 2 | 1081 | ||
XM_011528041.2 | XP_011526343.1 | 3 | 1185 | |||
GenBank mRNA | AL161995.1 | - | 2 | 1081 |