Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320924.1 | NP_001307853.1 | 2 | 600 | ||
NM_001320925.1 | NP_001307854.1 | 2 | 600 | |||
NM_032356.4 | NP_115732.2 | 1 | 784 | |||
XM_011524037.2 | XP_011522339.1 | 4 | 376 | |||
XM_017025225.1 | XP_016880714.1 | 4 | 283 | |||
XM_017025226.1 | XP_016880715.1 | 4 | 369 | |||
GenBank mRNA | AF530060.1 | - | 2 | 172 | ||
BC006407.2 | - | 2 | 271 | 3690202 | ||
BC033861.1 | - | 1 | 747 | 5171094 | ||
BC051846.1 | - | 2 | 600 | 4476019 | ||
BC059944.1 | - | 2 | 200 | 5223785 |