Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129993.1 | NP_001123465.1 | 20 | 2256 | ||
NM_032506.2 | NP_115895.2 | 20 | 2330 | |||
XM_005264605.3 | XP_005264662.1 | 20 | 2257 | |||
XM_005264606.4 | XP_005264663.1 | 20 | 2257 | |||
XM_005264607.2 | XP_005264664.1 | 19 | 2143 | |||
XM_011533126.2 | XP_011531428.1 | 20 | 2309 | |||
XM_011533127.2 | XP_011531429.1 | 20 | 2169 | |||
XM_011533128.1 | XP_011531430.1 | 20 | 2257 | |||
XM_011533129.2 | XP_011531431.1 | 19 | 2104 | |||
XM_017005108.1 | XP_016860597.1 | 20 | 2171 | |||
GenBank mRNA | AB058744.1 | - | 15 | 1368 | ||
AL833595.1 | - | 20 | 2196 | |||
BC039298.1 | - | 19 | 2153 | 5314819 | ||
BC070104.1 | - | 20 | 2330 | 5268265 |