Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000495.4 | NP_000486.1 | 48, 49 | 4964 | ||
NM_033380.2 | NP_203699.1 | 50, 51 | 4982 | |||
XM_011530849.2 | XP_011529151.2 | 51, 52 | 4730 | |||
XM_017029259.1 | XP_016884748.1 | 50, 51 | 4721 | |||
XM_017029260.1 | XP_016884749.1 | 49, 50 | 4712 | |||
XM_017029263.1 | XP_016884752.1 | 35, 36 | 3808 | |||
GenBank mRNA | AK294066.1 | - | 1, 2 | 76 | ||
AK307900.1 | - | 8, 9 | 1194 | |||
BC151846.1 | - | 50, 51 | 4944 | 8860442 | ||
KM282618.1 | - | 51, 52 | 4955 | |||
M31115.1 | - | 17, 18 | 1943 | |||
M58526.1 | - | 45, 46 | 4440 |