Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265612.1 | NP_001252541.1 | 16 | 2208 | ||
NM_016284.4 | NP_057368.3 | 16 | 2208 | |||
NM_206999.2 | NP_996882.1 | 16 | 2208 | |||
NR_049763.1 | 16 | 2208 | ||||
GenBank mRNA | AB023224.2 | - | 3 | 268 | ||
AK056927.1 | - | 7 | 743 | |||
AL832769.1 | - | 16 | 2157 | |||
AL833549.1 | - | 16 | 2168 | |||
BC040523.1 | - | 16 | 2160 | 5266600 | ||
BX537840.1 | - | 16 | 2154 | |||
BX647815.1 | - | 16 | 2148 | |||
CR749237.1 | - | 9 | 1148 | |||
EF553522.1 | - | 16 | 2157 |