Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144944.1 | NP_001138416.1 | 4 | 680 | ||
NM_001144945.1 | NP_001138417.1 | 4 | 602 | |||
NM_033546.3 | NP_291024.1 | 4 | 661 | |||
GenBank mRNA | AK291726.1 | - | 4 | 641 | ||
AV714583.1 | - | 5 | 588 | |||
AY320408.1 | - | 4 | 665 | |||
BC004994.1 | - | 4 | 580 | 2906108 | ||
BG696903.1 | - | 4 | 681 | |||
CN302451.1 | - | 4 | 583 | |||
D50372.1 | - | 4 | 614 | |||
D82058.1 | - | 4 | 594 | |||
U26162.1 | - | 4 | 635 |