Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021933.3 | NP_068752.2 | 7 | 1124 | ||
XM_005263487.3 | XP_005263544.1 | 7 | 953 | |||
XM_011541895.1 | XP_011540197.1 | 7 | 955 | |||
XM_011541896.1 | XP_011540198.1 | 7 | 848 | |||
GenBank mRNA | AK022500.1 | - | 7 | 904 | ||
AK024020.1 | - | 7 | 843 | |||
AK095928.1 | - | 3 | 721 | |||
BC008068.2 | - | 7 | 833 | 3050071 | ||
FJ618905.1 | - | 7 | 1124 |