Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256416.2 | NP_001243345.1 | 15 | 2472 | ||
NM_001256417.2 | NP_001243346.1 | 14 | 2298 | |||
NM_032264.4 | NP_115640.1 | 15 | 2508 | |||
NR_046176.2 | 18 | 3037 | ||||
XM_006710957.1 | XP_006711020.1 | 15 | 2366 | |||
XM_006710958.1 | XP_006711021.1 | 18 | 2822 | |||
XM_006710959.2 | XP_006711022.1 | 16 | 2606 | |||
XM_011542276.1 | XP_011540578.1 | 19 | 3079 | |||
XM_011542277.1 | XP_011540579.1 | 19 | 3064 | |||
XM_011542278.1 | XP_011540580.1 | 19 | 2893 | |||
XM_011542281.1 | XP_011540583.1 | 16 | 2429 | |||
XM_017002498.1 | XP_016857987.1 | 19 | 2878 | |||
XM_017002499.1 | XP_016857988.1 | 14 | 2144 | |||
XM_017002500.1 | XP_016857989.1 | 18 | 2897 | |||
XM_017002501.1 | XP_016857990.1 | 19 | 2782 | |||
XM_017002502.1 | XP_016857991.1 | 18 | 2711 | |||
GenBank mRNA | AK095602.1 | - | 18 | 2991 | ||
AK098438.1 | - | 5 | 753 | |||
AL136890.1 | - | 15 | 2285 | |||
AL834330.1 | - | 11 | 1481 | |||
BC024011.2 | - | 15 | 2399 | 4827023 |