Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM106CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026810.1 | 1 | 634 | |||
GenBank mRNA | BC098294.1 | - | 1 | 498 | ||
BC098363.1 | - | 1 | 498 | |||
DN989334.1 | - | 1 | 643 |
Gene Symbol: FAM106A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026809.1 | 1 | 519 | |||
GenBank mRNA | AK021862.1 | - | 1 | 519 | ||
BC098263.1 | - | 1 | 498 |
Gene Symbol: FAM106B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126160.1 | 1 | 634 |