Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERGIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015966.2 | NP_057050.1 | 6 | 602 | ||
NM_198398.1 | NP_938408.1 | 6 | 602 | |||
GenBank mRNA | AF077030.1 | - | 6 | 560 | ||
AF091085.1 | - | 4 | 213 | |||
AF116614.1 | - | 6 | 709 | |||
AF151812.1 | - | 6 | 560 | |||
AF308298.1 | - | 6 | 562 | |||
AK074550.1 | - | 6 | 550 | |||
AK300916.1 | - | 6 | 551 | |||
AL161963.1 | - | 3 | 243 | |||
BC009765.1 | - | 6 | 556 | 4283844 | ||
BC014014.2 | - | 6 | 555 | 3954269 | ||
BU528333.1 | - | 6 | 579 | |||
CA489637.1 | - | 6 | 602 | |||
EF206690.1 | - | 6 | 560 |