Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318323.1 | NP_001305252.1 | 5 | 693 | ||
NM_145912.6 | NP_666017.1 | 6 | 806 | |||
XM_005261374.4 | XP_005261431.1 | 8 | 1433 | |||
GenBank mRNA | AK074094.1 | - | 5 | 398 | ||
AY121370.1 | - | 6 | 806 | |||
AY247409.1 | - | 6 | 801 | |||
BC038241.1 | - | 6 | 801 | 5180790 | ||
CT841509.1 | - | 6 | 806 |