Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005776.2 | NP_005767.1 | 5 | 594 | ||
GenBank mRNA | AF070654.1 | - | 5 | 547 | ||
AF104398.1 | - | 5 | 547 | |||
AK074781.1 | - | 5 | 499 | |||
AK307074.1 | - | 4 | 355 | |||
AY358635.1 | - | 5 | 504 | |||
BC103741.1 | - | 5 | 595 | 6025795 | ||
BG503618.1 | - | 4 | 396 | |||
BQ937490.1 | - | 5 | 619 | |||
BX248284.1 | - | 4 | 343 |