Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193434.1 | NP_001180363.1 | 10 | 1315 | ||
NM_001193435.1 | NP_001180364.1 | 8 | 1119 | |||
NM_024889.4 | NP_079165.3 | 10 | 1215 | |||
NM_182601.1 | NP_872407.1 | 9 | 1432 | |||
XM_005270162.2 | XP_005270219.1 | 10 | 1354 | |||
XM_005270163.2 | XP_005270220.1 | 10 | 1354 | |||
XM_011540171.1 | XP_011538473.1 | 10 | 1354 | |||
XM_011540172.1 | XP_011538474.1 | 11 | 1122 | |||
XM_011540173.1 | XP_011538475.1 | 10 | 1354 | |||
XM_011540175.1 | XP_011538477.1 | 10 | 1293 | |||
XM_011540176.1 | XP_011538478.1 | 8 | 891 | |||
XM_017016666.1 | XP_016872155.1 | 10 | 1354 | |||
GenBank mRNA | AK027190.1 | - | 3 | 278 | ||
AK126354.1 | - | 8 | 891 | |||
AK128811.1 | - | 10 | 1215 | |||
AK292867.1 | - | 10 | 1087 | |||
BC036365.1 | - | 4 | 369 | |||
BC037870.1 | - | 9 | 1437 | 5273287 | ||
FJ236306.1 | - | 9 | 1437 |