Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193434.1 | NP_001180363.1 | 13 | 1719 | ||
NM_001193435.1 | NP_001180364.1 | 11 | 1523 | |||
NM_024889.4 | NP_079165.3 | 12 | 1525 | |||
XM_005270162.2 | XP_005270219.1 | 13 | 1800 | |||
XM_005270163.2 | XP_005270220.1 | 13 | 1758 | |||
GenBank mRNA | AK027190.1 | - | 6 | 682 | ||
AK126354.1 | - | 11 | 1295 | |||
AK128811.1 | - | 12 | 1525 | |||
AK292867.1 | - | 13 | 1491 | |||
AK296132.1 | - | 3 | 655 | |||
BC036365.1 | - | 7 | 815 |