Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM134A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321109.1 | NP_001308038.1 | 9 | 2433 | ||
NM_001321110.1 | NP_001308039.1 | 9 | 2185 | |||
NM_024293.5 | NP_077269.3 | 9 | 2521 | |||
XM_005246848.2 | XP_005246905.1 | 10 | 2389 | |||
GenBank mRNA | AK074983.1 | - | 9 | 2480 | ||
AK091011.1 | - | 10 | 2012 | |||
AL136758.2 | - | 1 | 1068 | |||
AL832843.1 | - | 3 | 1525 | |||
AL833630.1 | - | 3 | 1336 | |||
BC064950.1 | - | 3 | 1577 | |||
BC141851.1 | - | 9 | 2396 |