Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136037.2 | NP_001129509.2 | 10, 11 | 1103 | ||
NM_001161403.1 | NP_001154875.1 | 9, 10 | 1037 | |||
NM_001161404.1 | NP_001154876.1 | 9, 10 | 1327 | |||
NM_001256542.1 | NP_001243471.1 | 6, 7 | 729 | |||
NM_017980.4 | NP_060450.2 | 9, 10 | 1148 | |||
XM_005263710.2 | XP_005263767.1 | 8, 9 | 820 | |||
XM_006712627.3 | XP_006712690.1 | 6, 7 | 629 | |||
XM_011511453.1 | XP_011509755.1 | 8, 9 | 884 | |||
XM_017004469.1 | XP_016859958.1 | 4, 5 | 594 | |||
GenBank mRNA | AF520987.1 | - | 9, 10 | 1492 | ||
AF527764.1 | - | 9, 10 | 1102 | |||
AF527765.1 | - | 9, 10 | 964 | |||
AF527766.1 | - | 9, 10 | 1004 | |||
AF527767.1 | - | 8, 9 | 1260 | |||
AF527768.1 | - | 7, 8 | 619 | |||
AF527769.1 | - | 5, 6 | 693 | |||
AF527770.1 | - | 6, 7 | 656 | |||
AK000906.1 | - | 7, 8 | 519 | |||
AK055363.1 | - | 9, 10 | 1492 | |||
AK091652.1 | - | 7, 8 | 1398 | |||
AK093692.1 | - | 3, 4 | 1091 | |||
AK094954.1 | - | 8, 9 | 1584 | |||
AK095790.1 | - | 12, 13 | 1552 | |||
AK123014.1 | - | 4, 5 | 3796 | |||
AK297645.1 | - | 11, 12 | 1295 | |||
BC008977.1 | - | 1, 2 | 42 | |||
BC065816.1 | - | 9, 10 | 955 | 6181259 | ||
BX458594.2 | - | 6, 7 | 685 |