Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032315.2 | NP_115691.1 | 7 | 1292 | ||
XM_005263503.3 | XP_005263560.1 | 7 | 961 | |||
XM_011542296.1 | XP_011540598.1 | 7 | 1079 | |||
GenBank mRNA | AF495714.1 | - | 7 | 1260 | ||
AJ880283.1 | - | 7 | 1278 | |||
BC004991.1 | - | 7 | 1262 | 2905957 | ||
BC073135.1 | - | 7 | 1278 | 6172562 |