Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPHLN1 SIRNA Assays ›
Gene Symbol:
PPHLN1
Gene Name:
periphilin 1
Gene Aliases:
CR, HSPC206, HSPC232
Chromosome Location:
Chr.12: 42326126 - 42448621 on Build GRCh38
UniGene ID:
Hs.444157
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
215899
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPHLN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143787.1
NP_001137259.1
3
229
NM_001143788.1
NP_001137260.1
4
394
NM_001143789.1
NP_001137261.1
3
229
NM_016488.6
NP_057572.5
4
394
NM_201438.1
NP_958846.1
4
272
NM_201439.1
NP_958847.1
4
394
NM_201440.1
NP_958848.1
4
272
NM_201515.1
NP_958923.1
5
437
XM_005268962.3
XP_005269019.1
3
248
XM_011538459.2
XP_011536761.1
5
415
XM_011538465.2
XP_011536767.1
4
372
XM_011538467.2
XP_011536769.1
4
254
XM_011538470.2
XP_011536772.1
4
272
XM_011538471.2
XP_011536773.1
3
248
XM_017019434.1
XP_016874923.1
5
415
XM_017019435.1
XP_016874924.1
5
414
XM_017019436.1
XP_016874925.1
5
415
XM_017019437.1
XP_016874926.1
4
372
XM_017019438.1
XP_016874927.1
4
372
XM_017019439.1
XP_016874928.1
5
415
XM_017019440.1
XP_016874929.1
5
415
XM_017019441.1
XP_016874930.1
4
372
XM_017019442.1
XP_016874931.1
4
372
XM_017019443.1
XP_016874932.1
5
415
XM_017019444.1
XP_016874933.1
4
254
XM_017019445.1
XP_016874934.1
3
248
XM_017019446.1
XP_016874935.1
3
247
XM_017019447.1
XP_016874936.1
4
272
XM_017019448.1
XP_016874937.1
4
272
XM_017019449.1
XP_016874938.1
3
248
XM_017019450.1
XP_016874939.1
3
248
XM_017019451.1
XP_016874940.1
4
272
XM_017019452.1
XP_016874941.1
5
415
XM_017019453.1
XP_016874942.1
4
272
GenBank mRNA
AF151066.1
-
4
247
AK055690.1
-
4
382
AK129910.1
-
5
437
AK299951.1
-
3
211
AK303612.1
-
3
229
AK308022.1
-
4
368
AY039238.1
-
4
376
AY157850.1
-
5
433
BC025306.1
-
3
196
BC039832.1
-
4
239
6051282
BC040449.1
-
4
239
CF242917.1
-
3
244
DC424443.1
-
3
214
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
keratinization
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-cp9cp:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline