Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173689.6 | NP_775960.4 | 8 | 2109 | ||
NR_104603.1 | 3 | 1129 | ||||
XM_005251934.2 | XP_005251991.1 | 3 | 1129 | |||
XM_011518556.2 | XP_011516858.1 | 8 | 2109 | |||
XM_011518557.2 | XP_011516859.1 | 8 | 2111 | |||
XM_011518558.2 | XP_011516860.1 | 9 | 2471 | |||
GenBank mRNA | AK123000.1 | - | 8 | 2099 | ||
AK126775.1 | - | 3 | 1129 | |||
AY720432.1 | - | 8 | 2015 |