Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173689.6 | NP_775960.4 | 7 | 1541 | ||
NR_104603.1 | 2 | 561 | ||||
XM_005251934.2 | XP_005251991.1 | 2 | 561 | |||
XM_011518556.2 | XP_011516858.1 | 7 | 1541 | |||
XM_011518557.2 | XP_011516859.1 | 7 | 1543 | |||
XM_011518558.2 | XP_011516860.1 | 8 | 1903 | |||
GenBank mRNA | AK123000.1 | - | 7 | 1531 | ||
AK126775.1 | - | 2 | 561 | |||
AY720432.1 | - | 7 | 1447 |