Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145886.3 | NP_665893.2 | 5 | 1080 | ||
NM_145887.3 | NP_665894.2 | 5 | 1080 | |||
XM_005253005.4 | XP_005253062.1 | 6 | 2018 | |||
XM_005253006.4 | XP_005253063.1 | 6 | 1581 | |||
XM_005253007.3 | XP_005253064.1 | 5 | 1098 | |||
XM_005253008.4 | XP_005253065.1 | 6 | 2078 | |||
XM_011520209.2 | XP_011518511.1 | 5 | 1316 | |||
XM_011520210.2 | XP_011518512.1 | 6 | 2018 | |||
XM_011520211.2 | XP_011518513.1 | 6 | 2018 | |||
XM_011520212.1 | XP_011518514.1 | 5 | 1158 | |||
XM_011520213.2 | XP_011518515.1 | 3 | 300 | |||
XM_017017993.1 | XP_016873482.1 | 2 | 512 | |||
GenBank mRNA | AB208832.1 | - | 3 | 1884 | ||
AB208949.1 | - | 3 | 1848 | |||
AB209529.1 | - | 3 | 1990 | |||
AF229178.1 | - | 4 | 1282 | |||
AF274972.1 | - | 5 | 1076 | |||
AF465246.1 | - | 2 | 460 | |||
AK074893.1 | - | 2 | 83 | |||
AL833849.1 | - | 2 | 316 | |||
BC014904.1 | - | 4 | 1018 | 3908102 | ||
BQ924290.1 | - | 2 | 353 | |||
BX349115.2 | - | 3 | 433 |