Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003675.3 | NP_003666.1 | 10 | 1232 | ||
XM_011519754.2 | XP_011518056.1 | 11 | 1357 | |||
XM_017016860.1 | XP_016872349.1 | 9 | 1219 | |||
GenBank mRNA | AF087997.1 | - | 3 | 338 | ||
BC000794.1 | - | 10 | 1154 | 3451639 | ||
U51990.1 | - | 10 | 1156 |