Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318818.1 | NP_001305747.1 | 7 | 1283 | ||
NM_001318820.1 | NP_001305749.1 | 6 | 1341 | |||
NM_001318821.1 | NP_001305750.1 | 7 | 1449 | |||
NM_001318822.1 | NP_001305751.1 | 5 | 1264 | |||
NM_001318823.1 | NP_001305752.1 | 6 | 1372 | |||
NM_001318824.1 | NP_001305753.1 | 7 | 1252 | |||
NM_182612.3 | NP_872418.1 | 8 | 1360 | |||
NR_134867.1 | 7 | 1391 | ||||
NR_134868.1 | 8 | 1454 | ||||
XM_017017664.1 | XP_016873153.1 | 7 | 1475 | |||
XM_017017667.1 | XP_016873156.1 | 6 | 1381 | |||
XM_017017668.1 | XP_016873157.1 | 6 | 1463 | |||
GenBank mRNA | AK091602.1 | - | 8 | 1342 | ||
AK092816.1 | - | 8 | 957 | |||
AK094678.1 | - | 1 | 291 | |||
AK128653.1 | - | 5 | 1609 | |||
AK293559.1 | - | 7 | 1268 |