Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142885.1 | NP_001136357.1 | 3 | 454 | ||
NM_014548.3 | NP_055363.1 | 3 | 454 | |||
XM_017022089.1 | XP_016877578.1 | 3 | 418 | |||
GenBank mRNA | AF177169.1 | - | 3 | 389 | ||
AK292733.1 | - | 3 | 333 | |||
AK293823.1 | - | 3 | 342 | |||
BC036184.1 | - | 4 | 500 | 4815047 | ||
BC064961.1 | - | 3 | 402 | 6141541 | ||
DA808222.1 | - | 3 | 454 |