Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194247.2 | NP_919223.1 | 7 | 901 | ||
XM_005246380.1 | XP_005246437.1 | 7 | 901 | |||
XM_005246381.4 | XP_005246438.1 | 7 | 807 | |||
XM_006712365.2 | XP_006712428.1 | 7 | 901 | |||
XM_017003587.1 | XP_016859076.1 | 7 | 807 | |||
XM_017003588.1 | XP_016859077.1 | 7 | 901 | |||
GenBank mRNA | AK293125.1 | - | 7 | 769 | ||
AK316434.1 | - | 7 | 769 | |||
AL832246.1 | - | 1 | 66 | |||
AY363225.1 | - | 7 | 795 | |||
BC080591.1 | - | 3 | 229 | |||
BC112027.1 | - | 7 | 823 | 8327495 | ||
BC113470.1 | - | 7 | 823 | 8322522 |