Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301738.1 | NP_001288667.1 | 6 | 1254 | ||
NM_198282.3 | NP_938023.1 | 8 | 1682 | |||
XM_005268445.3 | XP_005268502.1 | 7 | 1425 | |||
XM_011537640.2 | XP_011535942.1 | 7 | 1385 | |||
GenBank mRNA | AK095896.1 | - | 6 | 2681 | ||
AK129800.1 | - | 2 | 536 | |||
AK290661.1 | - | 8 | 1619 | |||
AX775913.1 | - | 8 | 1590 | |||
BC047779.1 | - | 8 | 1599 | 5762441 |