Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181780.3 | NP_861445.3 | 3 | 635 | ||
XM_011512447.2 | XP_011510749.1 | 4 | 747 | |||
XM_017005748.1 | XP_016861237.1 | 4 | 710 | |||
GenBank mRNA | AJ717664.1 | - | 3 | 432 | ||
AK131204.1 | - | 3 | 424 | |||
AK292692.1 | - | 3 | 614 | |||
AY606805.1 | - | 3 | 432 | |||
BC107092.1 | - | 3 | 430 |