Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006641.3 | NP_001006642.1 | 10 | 1743 | ||
NM_001006642.3 | NP_001006643.1 | 10 | 1988 | |||
NM_001265614.2 | NP_001252543.1 | 11 | 2024 | |||
NM_052901.4 | NP_443133.2 | 10 | 1606 | |||
NR_049766.2 | 10 | 1649 | ||||
XM_005251688.1 | XP_005251745.1 | 11 | 1779 | |||
XM_005251689.4 | XP_005251746.1 | 10 | 2002 | |||
XM_006716948.2 | XP_006717011.1 | 11 | 2180 | |||
XM_006716949.3 | XP_006717012.1 | 11 | 2040 | |||
GenBank mRNA | AB067483.1 | - | 11 | 2023 | ||
AJ619963.1 | - | 10 | 1907 | |||
AJ619989.1 | - | 10 | 1531 | |||
AJ619990.1 | - | 10 | 1626 | |||
AJ619991.1 | - | 10 | 1588 | |||
AJ619992.1 | - | 10 | 1574 | |||
AK290705.1 | - | 10 | 1579 | |||
AK290991.1 | - | 10 | 1743 | |||
AY358515.1 | - | 10 | 1581 | |||
BC005163.2 | - | 6 | 1040 | |||
BC044631.1 | - | 1 | 166 | |||
BC089448.1 | - | 10 | 1597 | 30558551 | ||
BC103930.1 | - | 10 | 1548 | 40008034 | ||
BC103931.1 | - | 10 | 1548 | 40008035 | ||
BC103932.1 | - | 10 | 1548 | 40008036 | ||
BC103933.1 | - | 10 | 1548 | 40008037 |