Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198075.3 | NP_932341.1 | 8 | 945 | ||
XM_005252775.2 | XP_005252832.1 | 8 | 1769 | |||
XM_005252776.2 | XP_005252833.1 | 8 | 1769 | |||
XM_006718132.2 | XP_006718195.1 | 8 | 1844 | |||
XM_006718133.2 | XP_006718196.1 | 7 | 652 | |||
XM_011519875.2 | XP_011518177.1 | 9 | 1184 | |||
XM_011519876.1 | XP_011518178.1 | 9 | 1840 | |||
XM_011519877.2 | XP_011518179.1 | 8 | 921 | |||
XM_017017167.1 | XP_016872656.1 | 9 | 1259 | |||
XM_017017168.1 | XP_016872657.1 | 9 | 1498 | |||
GenBank mRNA | AK024495.1 | - | 7 | 2412 | ||
BC016439.1 | - | 6 | 479 | 3447051 | ||
BC035937.1 | - | 8 | 931 | 4816105 |