Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL22A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152888.2 | NP_690848.1 | 65 | 5932 | ||
XM_011516883.2 | XP_011515185.1 | 64 | 5950 | |||
XM_011516884.2 | XP_011515186.1 | 63 | 5911 | |||
XM_011516885.2 | XP_011515187.1 | 63 | 5896 | |||
XM_011516886.2 | XP_011515188.1 | 65 | 5862 | |||
XM_011516887.2 | XP_011515189.1 | 61 | 4830 | |||
XM_011516889.2 | XP_011515191.1 | 54 | 4071 | |||
XM_017013150.1 | XP_016868639.1 | 65 | 5823 | |||
XM_017013151.1 | XP_016868640.1 | 61 | 5788 | |||
XM_017013152.1 | XP_016868641.1 | 60 | 4820 | |||
GenBank mRNA | AF406780.1 | - | 65 | 5919 | ||
BC042075.1 | - | 39 | 3156 |