Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170714.1 | NP_001164185.1 | 3 | 958 | ||
NM_001170715.1 | NP_001164186.1 | 2 | 663 | |||
NM_001170716.1 | NP_001164187.1 | 2 | 620 | |||
NM_001170717.1 | NP_001164188.1 | 2 | 682 | |||
NM_001170718.1 | NP_001164189.1 | 2 | 707 | |||
NM_001170719.1 | NP_001164190.1 | 2 | 797 | |||
NM_014567.3 | NP_055382.2 | 2 | 682 | |||
XM_005256258.4 | XP_005256315.1 | 2 | 882 | |||
XM_005256259.4 | XP_005256316.1 | 2 | 3794 | |||
XM_005256260.4 | XP_005256317.1 | 2 | 803 | |||
XM_011523475.2 | XP_011521777.1 | 2 | 831 | |||
XM_017023897.1 | XP_016879386.1 | 2 | 1389 | |||
XM_017023898.1 | XP_016879387.1 | 2 | 1389 | |||
XM_017023899.1 | XP_016879388.1 | 2 | 1389 | |||
GenBank mRNA | AB040024.1 | - | 2 | 677 | ||
AF218451.1 | - | 2 | 660 | |||
AJ242987.2 | - | 2 | 657 | |||
AK027608.1 | - | 2 | 707 | |||
AK074958.1 | - | 1 | 92 | |||
AK124526.1 | - | 2 | 2102 | |||
AK124815.1 | - | 2 | 666 | |||
AK293808.1 | - | 3 | 958 | |||
AK294513.1 | - | 2 | 713 | |||
AK295809.1 | - | 2 | 592 | |||
AK314722.1 | - | 2 | 682 | |||
AY545071.1 | - | 1 | 536 | |||
BC062556.1 | - | 1 | 537 | 6427808 |