Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014225.5 | NP_055040.2 | 4, 5 | 792 | ||
NR_033500.1 | 3, 4 | 691 | ||||
XM_017026929.1 | XP_016882418.1 | 4, 5 | 1021 | |||
GenBank mRNA | AK090488.1 | - | 4, 5 | 1026 | ||
AK091387.1 | - | 4, 5 | 396 | |||
AK290593.1 | - | 4, 5 | 580 | |||
AK291932.1 | - | 4, 5 | 575 | |||
AK293174.1 | - | 5, 6 | 543 | |||
AK293493.1 | - | 3, 4 | 416 | |||
AK299012.1 | - | 4, 5 | 447 | |||
AK303934.1 | - | 893 | ||||
AK307743.1 | - | 4, 5 | 663 | |||
AK315700.1 | - | 4, 5 | 608 | |||
BC001537.2 | - | 4, 5 | 538 | 2987938 | ||
BC026733.1 | - | 4, 5 | 555 | |||
CR450340.1 | - | 4, 5 | 497 | |||
J02902.1 | - | 4, 5 | 641 |