Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCHIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197107.1 | NP_001184036.1 | 3 | 1372 | ||
NM_001197108.1 | NP_001184037.1 | 2 | 715 | |||
NM_001197109.1 | NP_001184038.1 | 2 | 432 | |||
NM_014575.3 | NP_055390.1 | 3 | 1411 | |||
GenBank mRNA | AF070614.1 | - | 2 | 521 | ||
AF145713.1 | - | 3 | 906 | |||
AF145714.1 | - | 2 | 210 | |||
AF145715.1 | - | 3 | 867 | |||
AK091871.1 | - | 2 | 353 | |||
AK309223.1 | - | 3 | 819 | |||
AK314662.1 | - | 3 | 1354 | |||
BC005947.1 | - | 2 | 248 | 4246326 | ||
BC036535.2 | - | 3 | 1411 | 5263291 | ||
BX648179.1 | - | 2 | 546 |
Gene Symbol: IQCJ-SCHIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197113.1 | NP_001184042.1 | 6 | 1246 | ||
NM_001197114.1 | NP_001184043.1 | 5 | 1165 | |||
GenBank mRNA | AK096479.1 | - | 6 | 1152 | ||
DQ157847.1 | - | 4 | 984 | |||
DQ157848.1 | - | 5 | 1065 |