Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024091.3 | NP_076996.2 | 6 | 2034 | ||
NR_036553.1 | 5 | 483 | ||||
NR_073608.1 | 5 | 604 | ||||
XM_006714498.1 | XP_006714561.1 | 6 | 1945 | |||
GenBank mRNA | AF161354.1 | - | 5 | 457 | ||
AK026927.1 | - | 6 | 1999 | |||
BC001295.1 | - | 5 | 1526 | |||
BC113563.1 | - | 5 | 1949 | 8322615 | ||
BC143517.1 | - | 5 | 2070 | |||
BC143518.1 | - | 6 | 1386 | |||
BG613096.1 | - | 5 | 595 |