Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130820.2 | NP_001124292.1 | 4 | 847 | ||
NM_001130821.2 | NP_001124293.1 | 5 | 968 | |||
NM_001130822.2 | NP_001124294.1 | 4 | 860 | |||
NM_001287525.1 | NP_001274454.1 | 6 | 1010 | |||
NM_001287526.1 | NP_001274455.1 | 3 | 770 | |||
NM_022777.3 | NP_073614.1 | 5 | 937 | |||
GenBank mRNA | AK023287.1 | - | 6 | 968 | ||
AK024179.1 | - | 5 | 863 | |||
AK298888.1 | - | 4 | 801 | |||
AL157469.1 | - | 2 | 549 | |||
BC004522.2 | - | 4 | 801 | 3939191 | ||
BC009823.2 | - | 5 | 910 | 3946565 | ||
BC038668.1 | - | 4 | 1647 | 5261016 |