Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130820.2 | NP_001124292.1 | 4 | 532 | ||
NM_001130821.2 | NP_001124293.1 | 5 | 653 | |||
NM_001130822.2 | NP_001124294.1 | 4 | 545 | |||
NM_001287525.1 | NP_001274454.1 | 6 | 695 | |||
NM_001287526.1 | NP_001274455.1 | 3 | 455 | |||
NM_022777.3 | NP_073614.1 | 5 | 622 | |||
GenBank mRNA | AK023287.1 | - | 6 | 653 | ||
AK024179.1 | - | 5 | 548 | |||
AK298888.1 | - | 4 | 486 | |||
AL157469.1 | - | 2 | 234 | |||
BC004522.2 | - | 4 | 486 | 3939191 | ||
BC009823.2 | - | 5 | 595 | 3946565 | ||
BC038668.1 | - | 4 | 1332 | 5261016 | ||
BU597007.1 | - | 5 | 631 | |||
DA700693.1 | - | 3 | 381 | |||
DA709553.1 | - | 3 | 458 | |||
DN993868.1 | - | 5 | 631 |