Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032888.3 | NP_116277.2 | 3 | 1931 | ||
XM_006717308.2 | XP_006717371.1 | 3 | 1999 | |||
XM_011519138.2 | XP_011517440.1 | 3 | 1563 | |||
XM_011519140.1 | XP_011517442.1 | 3 | 1999 | |||
XM_011519142.2 | XP_011517444.1 | 3 | 2005 | |||
XM_011519143.2 | XP_011517445.1 | 3 | 2003 | |||
XM_011519144.2 | XP_011517446.1 | 3 | 2003 | |||
XM_017015239.1 | XP_016870728.1 | 3 | 2004 | |||
GenBank mRNA | AB058773.1 | - | 1 | 473 | ||
AY149237.1 | - | 3 | 1525 | |||
BC080610.1 | - | 1 | 814 |