Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032888.3 | NP_116277.2 | 58 | 5388 | ||
XM_006717308.2 | XP_006717371.1 | 57 | 5402 | |||
XM_006717310.3 | XP_006717373.1 | 56 | 3201 | |||
XM_011519138.2 | XP_011517440.1 | 58 | 5020 | |||
XM_011519140.1 | XP_011517442.1 | 57 | 5402 | |||
XM_011519145.2 | XP_011517447.1 | 45 | 2630 | |||
GenBank mRNA | AB058773.1 | - | 55 | 3875 | ||
AY149237.1 | - | 58 | 4982 |