Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032888.3 | NP_116277.2 | 20, 21 | 3187 | ||
XM_006717308.2 | XP_006717371.1 | 19, 20 | 3201 | |||
XM_006717310.3 | XP_006717373.1 | 18, 19 | 1000 | |||
XM_011519138.2 | XP_011517440.1 | 20, 21 | 2819 | |||
XM_011519140.1 | XP_011517442.1 | 20, 21 | 3255 | |||
XM_011519142.2 | XP_011517444.1 | 20, 21 | 3261 | |||
XM_011519143.2 | XP_011517445.1 | 20, 21 | 3259 | |||
XM_011519144.2 | XP_011517446.1 | 20, 21 | 3259 | |||
XM_011519145.2 | XP_011517447.1 | 7, 8 | 429 | |||
XM_017015239.1 | XP_016870728.1 | 20, 21 | 3260 | |||
GenBank mRNA | AB058773.1 | - | 17, 18 | 1667 | ||
AY149237.1 | - | 20, 21 | 2781 |